Search
Search

CUNH1orf106| Gene Symbol: CUNH1orf106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862262.2 | XP_004862319.1 | 5 - 6 | 845 | 92 | |
| XM_021242422.1 | XP_021098081.1 | 5 - 6 | 818 | 92 | ||
| XM_021242424.1 | XP_021098083.1 | 5 - 6 | 747 | 92 | ||
Set Membership: |
|