Search
Search

Col12a1| Gene Symbol: Col12a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013065253.2 | XP_012920707.1 | 41 - 42 | 7195 | 72 | |
| XM_013065254.2 | XP_012920708.1 | 41 - 42 | 7194 | 72 | ||
| XM_021243751.1 | XP_021099410.1 | 26 - 27 | 3691 | 72 | ||
Set Membership: |
|