Search
Search

Zmiz1| Gene Symbol: Zmiz1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846383.3 | XP_004846440.1 | 18 - 19 | 2675 | 69 | |
| XM_004846384.3 | XP_004846441.1 | 17 - 18 | 2447 | 69 | ||
| XM_004846385.3 | XP_004846442.1 | 17 - 18 | 2447 | 69 | ||
| XM_004846386.3 | XP_004846443.1 | 18 - 19 | 2656 | 69 | ||
| XM_004846387.3 | XP_004846444.1 | 18 - 19 | 2679 | 69 | ||
| XM_021259686.1 | XP_021115345.1 | 17 - 18 | 2447 | 69 | ||
| XM_021259687.1 | XP_021115346.1 | 13 - 14 | 2229 | 69 | ||
Set Membership: |
|