Search
Search

Efcab11| Gene Symbol: Efcab11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837076.3 | XP_004837133.1 | 4 - 5 | 442 | 93 | |
| XM_004837077.2 | XP_004837134.1 | 4 - 5 | 442 | 93 | ||
| XM_021245235.1 | XP_021100894.1 | 4 - 5 | 446 | 93 | ||
| XM_021245240.1 | XP_021100899.1 | 4 - 5 | 442 | 93 | ||
| XM_021245246.1 | XP_021100905.1 | 4 - 5 | 444 | 93 | ||
| XM_021245258.1 | XP_021100917.1 | 4 - 5 | 369 | 93 | ||
| XM_021245261.1 | XP_021100920.1 | 4 - 5 | 532 | 93 | ||
Set Membership: |
|