Search
Search

Kif17| Gene Symbol: Kif17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021262961.1 | XP_021118620.1 | 2 - 3 | 650 | 61 | |
| XM_021262962.1 | XP_021118621.1 | 2 - 3 | 650 | 61 | ||
| XM_021262963.1 | XP_021118622.1 | 2 - 3 | 652 | 61 | ||
| XM_021262964.1 | XP_021118623.1 | 2 - 3 | 652 | 61 | ||
| XM_021262965.1 | XP_021118624.1 | 2 - 3 | 647 | 61 | ||
| XM_021262966.1 | XP_021118625.1 | 2 - 3 | 650 | 61 | ||
Set Membership: |
|