| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009439.hg.1 |
ENST00000390372.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009439.hg.1 |
ENST00000390372.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009436.hg.1 |
ENST00000454561.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009436.hg.1 |
ENST00000454561.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009426.hg.1 |
ENST00000390362.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009426.hg.1 |
ENST00000390362.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009445.hg.1 |
ENST00000390378.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009445.hg.1 |
ENST00000390378.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000390398.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000390398.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000390398.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000390398.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000390398.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000390399.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000390399.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000390399.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000390399.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000390399.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000390399.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000390400.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000390400.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000390400.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000390412.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000390416.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000390418.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000466254.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000466254.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000466254.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000466254.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000466254.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000466254.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000610416.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000610416.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000610416.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000610416.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000610416.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000610416.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000610416.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000610416.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000610566.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000610566.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000610566.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000610566.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000610566.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000610566.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000610566.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000610566.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000610566.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000610847.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000610847.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000610847.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000610847.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000610847.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000610847.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000610847.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000610847.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000610847.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000610847.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000612107.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000612107.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000612107.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000612107.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000612107.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000612107.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000612107.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000613761.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000613761.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000613761.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000613761.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000613761.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000613761.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000613761.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000613761.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000613761.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000613761.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000616837.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000616837.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000616837.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000616837.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000616837.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000616837.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000616837.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000616837.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000616837.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000617717.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000617717.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000617717.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000617717.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000617717.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000617717.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000617717.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000633705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000633705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000633705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000633705.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000633713.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013479.hg.1 |
ENST00000634000.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009442.hg.1 |
ENST00000390375.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700009442.hg.1 |
ENST00000390375.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009426.hg.1 |
ENST00000390362.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009426.hg.1 |
ENST00000390362.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009436.hg.1 |
ENST00000454561.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009436.hg.1 |
ENST00000454561.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009439.hg.1 |
ENST00000390372.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009439.hg.1 |
ENST00000390372.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009445.hg.1 |
ENST00000390378.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009445.hg.1 |
ENST00000390378.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000390398.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000390398.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000390398.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000390398.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000390398.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000390399.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000390399.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000390399.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000390399.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000390399.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000390399.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000390400.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000390400.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000390400.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000390412.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000390416.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000390418.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000466254.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000466254.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000466254.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000466254.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000466254.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000466254.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000610416.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000610416.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000610416.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000610416.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000610416.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000610416.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000610416.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000610416.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000610566.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000610566.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000610566.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000610566.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000610566.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000610566.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000610566.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000610566.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000610566.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000610847.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000610847.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000610847.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000610847.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000610847.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000610847.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000610847.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000610847.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000610847.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000610847.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000612107.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000612107.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000612107.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000612107.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000612107.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000612107.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000612107.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000613761.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000613761.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000613761.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000613761.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000613761.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000613761.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000613761.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000613761.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000613761.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000613761.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000616837.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000616837.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000616837.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000616837.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000616837.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000616837.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000616837.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000616837.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000616837.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000617717.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000617717.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000617717.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000617717.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000617717.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000617717.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000617717.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000633705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000633705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000633705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000633705.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000633713.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013479.hg.1 |
ENST00000634000.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009442.hg.1 |
ENST00000390375.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700009442.hg.1 |
ENST00000390375.2 |
Ensembl |