Search
Search
| Gene Symbol: NRP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017005186.1 | XP_016860675.1 | 17 | 5272 | 72 | |
| GenBank mRNA | AF119893.2 | - | 1 | 741 | 72 | |
| AK024680.1 | - | 1 | 622 | 72 | ||
Set Membership: |
|