Search
Search

CLCN1| Gene Symbol: CLCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000083.2 | NP_000074.2 | 20 - 21 | 2496 | 113 | |
| NR_046453.1 | 19 - 20 | 2349 | 113 | |||
| XM_011515782.2 | XP_011514084.1 | 10 - 11 | 1244 | 113 | ||
| XM_017011739.1 | XP_016867228.1 | 17 - 18 | 2221 | 113 | ||
| XM_017011740.1 | XP_016867229.1 | 17 - 18 | 2197 | 113 | ||
| GenBank mRNA | BC112156.1 | - | 20 - 21 | 2461 | 8327624 | 113 |
| BC113495.1 | - | 20 - 21 | 2461 | 8322547 | 113 | |
| BC143796.1 | - | 19 - 20 | 2314 | 113 | ||
| M97820.1 | - | 17 - 18 | 1900 | 113 | ||
| Z25884.1 | - | 20 - 21 | 2496 | 113 | ||
Set Membership: |
|