| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008786.hg.1 |
ENST00000217289.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008786.hg.1 |
ENST00000339538.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008786.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008786.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008786.hg.1 |
TC20000603.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008793.hg.1 |
ENST00000217289.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008793.hg.1 |
ENST00000339538.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008793.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008793.hg.1 |
TC20000603.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000378844.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000378844.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000378844.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000378844.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008794.hg.1 |
ENST00000217289.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008794.hg.1 |
ENST00000339538.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008794.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008794.hg.1 |
TC20000603.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008792.hg.1 |
ENST00000217289.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008792.hg.1 |
ENST00000339538.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008792.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008792.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008792.hg.1 |
TC20000603.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008789.hg.1 |
ENST00000217289.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008789.hg.1 |
ENST00000339538.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008789.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008789.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20008789.hg.1 |
TC20000603.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR2000158648.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158648.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000158648.hg.1 |
TC2000008318.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000158648.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158648.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000378844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000378844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000378844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000378844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000478194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000008318.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000158640.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158640.hg.1 |
ENST00000378844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158640.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158640.hg.1 |
NM_017671.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000158640.hg.1 |
TC2000008318.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000158640.hg.1 |
ENST00000217289.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158640.hg.1 |
ENST00000378844.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158640.hg.1 |
ENST00000536936.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000158640.hg.1 |
NM_017671.4 |
Refseq |