| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000359927.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000359927.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000359927.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09002551.hg.1 |
ENST00000377765.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09002551.hg.1 |
ENST00000536622.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09002551.hg.1 |
ENST00000539465.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09002551.hg.1 |
ENST00000541302.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09002551.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09002551.hg.1 |
TC09000216.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000359927.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000359927.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000359927.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900007163.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900148848.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900148848.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900148848.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900148848.hg.1 |
ENST00000377765.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900148848.hg.1 |
ENST00000539465.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900148848.hg.1 |
NM_014907.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900148848.hg.1 |
TC0900007163.hg.1 |
Clariom D TC |