Search
Search

SCNN1D| Gene Symbol: SCNN1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130413.3 | NP_001123885.2 | 12 - 13 | 1888 | 68 | |
| NR_037668.2 | 11 - 12 | 1794 | 68 | |||
| XM_011541899.2 | XP_011540201.1 | 12 - 13 | 2558 | 68 | ||
| XM_011541901.2 | XP_011540203.1 | 13 - 14 | 2080 | 68 | ||
| XM_011541902.2 | XP_011540204.1 | 13 - 14 | 2056 | 68 | ||
| XM_011541905.2 | XP_011540207.1 | 12 - 13 | 1984 | 68 | ||
| XM_011541906.2 | XP_011540208.1 | 9 - 10 | 2318 | 68 | ||
| XM_011541908.2 | XP_011540210.1 | 12 - 13 | 2255 | 68 | ||
| XM_011541920.2 | XP_011540222.1 | 11 - 12 | 2078 | 68 | ||
| XM_011541925.2 | XP_011540227.1 | 10 - 11 | 1637 | 68 | ||
| XM_011541929.2 | XP_011540231.1 | 8 - 9 | 2446 | 68 | ||
| XM_011541932.2 | XP_011540234.1 | 9 - 10 | 1543 | 68 | ||
| XM_011541933.2 | XP_011540235.1 | 9 - 10 | 1509 | 68 | ||
| XM_017002037.1 | XP_016857526.1 | 11 - 12 | 2555 | 68 | ||
| XM_017002038.1 | XP_016857527.1 | 10 - 11 | 2408 | 68 | ||
| XM_017002039.1 | XP_016857528.1 | 12 - 13 | 1933 | 68 | ||
| XM_017002040.1 | XP_016857529.1 | 12 - 13 | 1795 | 68 | ||
| XM_017002041.1 | XP_016857530.1 | 13 - 14 | 1783 | 68 | ||
| XM_017002042.1 | XP_016857531.1 | 12 - 13 | 1774 | 68 | ||
| XM_017002043.1 | XP_016857532.1 | 12 - 13 | 1750 | 68 | ||
| XM_017002044.1 | XP_016857533.1 | 11 - 12 | 1699 | 68 | ||
| GenBank mRNA | AK093239.1 | - | 9 - 10 | 1518 | 68 | |
| AK093372.1 | - | 6 - 7 | 643 | 68 | ||
| AK127357.1 | - | 9 - 10 | 2314 | 68 | ||
| AK131558.1 | - | 13 - 14 | 2214 | 68 | ||
| AK315288.1 | - | 8 - 9 | 1365 | 68 | ||
| BC036752.1 | - | 8 - 9 | 1337 | 68 | ||
| BC040882.1 | - | 11 - 12 | 1758 | 68 | ||
| BC125074.1 | - | 8 - 9 | 1287 | 40117641 | 68 | |
| BC125075.1 | - | 7 - 8 | 1389 | 40117643 | 68 | |
| DQ898175.1 | - | 8 - 9 | 1262 | 68 | ||
| DQ898176.1 | - | 9 - 10 | 1368 | 68 | ||
| EU489064.1 | - | 7 - 8 | 1170 | 68 | ||
| U38254.1 | - | 7 - 8 | 1170 | 68 | ||
Set Membership: |
|