Search
Search

BBOF1| Gene Symbol: BBOF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025057.2 | NP_079333.2 | 6 - 7 | 773 | 103 | |
| XM_005268092.3 | XP_005268149.1 | 6 - 7 | 779 | 103 | ||
| XM_011537170.2 | XP_011535472.1 | 6 - 7 | 779 | 103 | ||
| XM_011537171.2 | XP_011535473.1 | 6 - 7 | 779 | 103 | ||
| XM_011537172.1 | XP_011535474.1 | 6 - 7 | 779 | 103 | ||
| XM_011537174.2 | XP_011535476.1 | 6 - 7 | 779 | 103 | ||
| XM_011537175.2 | XP_011535477.1 | 5 - 6 | 635 | 103 | ||
| XM_011537176.2 | XP_011535478.1 | 7 - 8 | 763 | 103 | ||
| XM_011537177.2 | XP_011535479.1 | 5 - 6 | 483 | 103 | ||
| XM_011537178.2 | XP_011535480.1 | 5 - 6 | 454 | 103 | ||
| XM_011537179.2 | XP_011535481.1 | 4 - 5 | 339 | 103 | ||
| XM_017021659.1 | XP_016877148.1 | 6 - 7 | 779 | 103 | ||
| XM_017021660.1 | XP_016877149.1 | 5 - 6 | 495 | 103 | ||
| XM_017021661.1 | XP_016877150.1 | 5 - 6 | 547 | 103 | ||
| XM_017021662.1 | XP_016877151.1 | 4 - 5 | 331 | 103 | ||
| GenBank mRNA | AL834487.1 | - | 1 - 2 | 48 | 103 | |
Set Membership: |
|