| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008072.hg.1 |
ENST00000259782.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008072.hg.1 |
ENST00000339741.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008072.hg.1 |
ENST00000370864.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008072.hg.1 |
ENST00000370865.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008072.hg.1 |
ENST00000370869.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008072.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008072.hg.1 |
TC06000672.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008071.hg.1 |
ENST00000259782.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008071.hg.1 |
ENST00000339741.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008071.hg.1 |
ENST00000370864.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008071.hg.1 |
ENST00000370865.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008071.hg.1 |
ENST00000370869.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008071.hg.1 |
ENST00000486436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008071.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008071.hg.1 |
TC06000672.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
ENST00000259782.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
ENST00000259782.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
ENST00000259782.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
ENST00000259782.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
ENST00000259782.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
ENST00000259782.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
ENST00000259782.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
ENST00000259782.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
ENST00000259782.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
ENST00000259782.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
ENST00000259782.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
ENST00000259782.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
ENST00000259782.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
ENST00000259782.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
ENST00000259782.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
ENST00000370864.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
ENST00000370864.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
ENST00000370864.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
ENST00000370864.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
ENST00000370864.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
ENST00000370864.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
ENST00000370869.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
ENST00000370869.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
ENST00000370869.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
ENST00000370869.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
ENST00000370869.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
ENST00000370869.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
ENST00000370869.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
ENST00000486436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
ENST00000486436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
ENST00000486436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
ENST00000486436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
ENST00000486436.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008310.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600158041.hg.1 |
ENST00000259782.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600158041.hg.1 |
ENST00000370864.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600158041.hg.1 |
ENST00000370869.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600158041.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600158041.hg.1 |
ENST00000259782.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600158041.hg.1 |
ENST00000370864.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600158041.hg.1 |
ENST00000370869.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600158041.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600158041.hg.1 |
TC0600008310.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600158040.hg.1 |
ENST00000259782.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600158040.hg.1 |
ENST00000370864.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600158040.hg.1 |
ENST00000370869.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600158040.hg.1 |
ENST00000486436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600158040.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600158040.hg.1 |
ENST00000259782.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600158040.hg.1 |
ENST00000370864.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600158040.hg.1 |
ENST00000370869.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600158040.hg.1 |
ENST00000486436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600158040.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600158040.hg.1 |
TC0600008310.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
ENST00000259782.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
ENST00000259782.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
ENST00000259782.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
ENST00000259782.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
ENST00000259782.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
ENST00000259782.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
ENST00000259782.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
ENST00000259782.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
ENST00000259782.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
ENST00000259782.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
ENST00000259782.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
ENST00000259782.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
ENST00000259782.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
ENST00000259782.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
ENST00000259782.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
ENST00000370864.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
ENST00000370864.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
ENST00000370864.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
ENST00000370864.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
ENST00000370864.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
ENST00000370864.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
ENST00000370869.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
ENST00000370869.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
ENST00000370869.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
ENST00000370869.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
ENST00000370869.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
ENST00000370869.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
ENST00000370869.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
ENST00000486436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
ENST00000486436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
ENST00000486436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
ENST00000486436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
ENST00000486436.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
NM_014464.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008310.hg.1 |
NM_014464.3 |
Refseq |