| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08011634.hg.1 |
ENST00000262109.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08011634.hg.1 |
ENST00000522706.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08011634.hg.1 |
ENST00000543819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08011634.hg.1 |
NM_207332.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08011634.hg.1 |
TC08000881.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08011637.hg.1 |
ENST00000262109.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08011637.hg.1 |
ENST00000522706.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08011637.hg.1 |
ENST00000543819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08011637.hg.1 |
NM_207332.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08011637.hg.1 |
TC08000881.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08011638.hg.1 |
ENST00000262109.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08011638.hg.1 |
ENST00000522706.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08011638.hg.1 |
ENST00000543819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08011638.hg.1 |
NM_207332.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08011638.hg.1 |
TC08000881.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0800163935.hg.1 |
ENST00000519909.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163935.hg.1 |
ENST00000523415.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163935.hg.1 |
NM_001303100.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800163935.hg.1 |
ENST00000519909.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163935.hg.1 |
ENST00000523415.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163935.hg.1 |
NM_001303100.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800163935.hg.1 |
TC0800009331.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800163972.hg.1 |
ENST00000262109.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163972.hg.1 |
ENST00000518895.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163972.hg.1 |
ENST00000522706.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163972.hg.1 |
ENST00000522893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163972.hg.1 |
ENST00000523415.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163972.hg.1 |
NM_001303100.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800163972.hg.1 |
NM_207332.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800163972.hg.1 |
ENST00000262109.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163972.hg.1 |
ENST00000518895.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163972.hg.1 |
ENST00000522706.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163972.hg.1 |
ENST00000522893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163972.hg.1 |
ENST00000523415.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163972.hg.1 |
NM_001303100.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800163972.hg.1 |
NM_207332.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800163972.hg.1 |
TC0800009331.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800163932.hg.1 |
ENST00000523415.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163932.hg.1 |
NM_001303100.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800163932.hg.1 |
ENST00000523415.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163932.hg.1 |
NM_001303100.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800163932.hg.1 |
TC0800009331.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800163933.hg.1 |
ENST00000523415.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163933.hg.1 |
NM_001303100.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800163933.hg.1 |
ENST00000523415.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163933.hg.1 |
NM_001303100.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800163933.hg.1 |
TC0800009331.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800163930.hg.1 |
ENST00000522706.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163930.hg.1 |
ENST00000523415.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163930.hg.1 |
NM_001303100.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800163930.hg.1 |
ENST00000522706.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163930.hg.1 |
ENST00000523415.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163930.hg.1 |
NM_001303100.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800163930.hg.1 |
TC0800009331.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800163963.hg.1 |
ENST00000262109.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163963.hg.1 |
ENST00000522706.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163963.hg.1 |
ENST00000522893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163963.hg.1 |
ENST00000523053.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163963.hg.1 |
ENST00000523415.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163963.hg.1 |
NM_001303100.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800163963.hg.1 |
NM_207332.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800163963.hg.1 |
ENST00000262109.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163963.hg.1 |
ENST00000522706.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163963.hg.1 |
ENST00000522893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163963.hg.1 |
ENST00000523053.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163963.hg.1 |
ENST00000523415.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163963.hg.1 |
NM_001303100.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800163963.hg.1 |
NM_207332.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800163963.hg.1 |
TC0800009331.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800163973.hg.1 |
ENST00000262109.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163973.hg.1 |
ENST00000518895.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163973.hg.1 |
ENST00000522706.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163973.hg.1 |
ENST00000522893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163973.hg.1 |
NM_001303100.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800163973.hg.1 |
NM_207332.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800163973.hg.1 |
ENST00000262109.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163973.hg.1 |
ENST00000518895.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163973.hg.1 |
ENST00000522706.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163973.hg.1 |
ENST00000522893.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163973.hg.1 |
NM_001303100.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800163973.hg.1 |
NM_207332.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800163973.hg.1 |
TC0800009331.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800163931.hg.1 |
ENST00000523415.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163931.hg.1 |
NM_001303100.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800163931.hg.1 |
ENST00000523415.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163931.hg.1 |
NM_001303100.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800163931.hg.1 |
TC0800009331.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0800163934.hg.1 |
ENST00000523415.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163934.hg.1 |
NM_001303100.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800163934.hg.1 |
ENST00000523415.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800163934.hg.1 |
NM_001303100.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800163934.hg.1 |
TC0800009331.hg.1 |
Clariom D TC |