| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06022073.hg.1 |
ENST00000244728.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06022073.hg.1 |
ENST00000370808.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06022073.hg.1 |
ENST00000370811.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06022073.hg.1 |
ENST00000370819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06022073.hg.1 |
ENST00000467045.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06022073.hg.1 |
ENST00000535941.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06022073.hg.1 |
NM_030820.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06022073.hg.1 |
TC06001826.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0600183436.hg.1 |
ENST00000244728.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183436.hg.1 |
ENST00000370819.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183436.hg.1 |
ENST00000461489.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183436.hg.1 |
ENST00000467045.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183436.hg.1 |
ENST00000484987.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183436.hg.1 |
ENST00000488912.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183436.hg.1 |
ENST00000535941.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183436.hg.1 |
NM_030820.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600183436.hg.1 |
ENST00000244728.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183436.hg.1 |
ENST00000370819.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183436.hg.1 |
ENST00000461489.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183436.hg.1 |
ENST00000467045.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183436.hg.1 |
ENST00000484987.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183436.hg.1 |
ENST00000488912.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183436.hg.1 |
ENST00000535941.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600183436.hg.1 |
NM_030820.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600183436.hg.1 |
TC0600012116.hg.1 |
Clariom D TC |