| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012058.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22012058.hg.1 |
TC22000682.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000463455.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000463455.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000463455.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000463455.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000463455.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000476619.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000476619.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000476619.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000476619.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000476619.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000485373.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000485373.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000485373.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000485373.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000485373.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000487704.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000487704.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000487704.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000487704.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000487704.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000487704.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000487704.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000498434.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000498434.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000498434.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
ENST00000498434.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200008514.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200161325.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200161325.hg.1 |
TC2200008514.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200161325.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000463455.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000463455.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000463455.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000463455.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000463455.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000476619.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000476619.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000476619.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000476619.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000476619.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000485373.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000485373.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000485373.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000485373.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000485373.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000487704.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000487704.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000487704.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000487704.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000487704.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000487704.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000487704.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000498434.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000498434.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000498434.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
ENST00000498434.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200008514.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200161317.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200161317.hg.1 |
ENST00000463455.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200161317.hg.1 |
ENST00000487704.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200161317.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200161317.hg.1 |
TC2200008514.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200161317.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200161317.hg.1 |
ENST00000463455.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200161317.hg.1 |
ENST00000487704.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200161317.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200161318.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200161318.hg.1 |
ENST00000463455.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200161318.hg.1 |
ENST00000487704.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200161318.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200161318.hg.1 |
TC2200008514.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200161318.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200161318.hg.1 |
ENST00000463455.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200161318.hg.1 |
ENST00000487704.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200161318.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200161319.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200161319.hg.1 |
ENST00000463455.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200161319.hg.1 |
ENST00000487704.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200161319.hg.1 |
NM_014306.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200161319.hg.1 |
TC2200008514.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200161319.hg.1 |
ENST00000216038.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200161319.hg.1 |
ENST00000463455.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200161319.hg.1 |
ENST00000487704.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200161319.hg.1 |
NM_014306.4 |
Refseq |