| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04006205.hg.1 |
ENST00000264895.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04006205.hg.1 |
ENST00000325942.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04006205.hg.1 |
NM_001166133.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04006205.hg.1 |
NM_025074.6 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04006205.hg.1 |
TC04000434.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0400154476.hg.1 |
ENST00000325942.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400154476.hg.1 |
ENST00000512123.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400154476.hg.1 |
NM_001166133.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400154476.hg.1 |
NM_025074.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400154476.hg.1 |
ENST00000325942.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400154476.hg.1 |
ENST00000512123.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400154476.hg.1 |
NM_001166133.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400154476.hg.1 |
NM_025074.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400154476.hg.1 |
TC0400007928.hg.1 |
Clariom D TC |