| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000444777.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000444777.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000444777.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000468591.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000468591.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000468591.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000468591.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000468591.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000468591.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000487069.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000487069.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000487069.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000487069.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000487069.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000487069.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000534978.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000534978.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000534978.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000534978.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000534978.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000534978.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000534978.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000534978.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000534978.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000534978.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000534978.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000534978.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
ENST00000534978.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
NM_030636.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
NM_030636.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
NM_030636.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
NM_030636.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
NM_030636.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
NM_030636.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
NM_030636.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
NM_030636.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
NM_030636.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
NM_030636.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
NM_030636.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
NM_030636.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
NM_030636.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
NM_030636.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
NM_030636.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700007191.hg.1 |
NM_030636.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07003598.hg.1 |
ENST00000242108.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07003598.hg.1 |
ENST00000534978.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07003598.hg.1 |
NM_030636.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07003598.hg.1 |
TC07000234.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0700150059.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700150059.hg.1 |
ENST00000468591.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700150059.hg.1 |
ENST00000487069.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700150059.hg.1 |
ENST00000534978.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700150059.hg.1 |
NM_030636.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700150059.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700150059.hg.1 |
ENST00000468591.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700150059.hg.1 |
ENST00000487069.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700150059.hg.1 |
ENST00000534978.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700150059.hg.1 |
NM_030636.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700150059.hg.1 |
TC0700007191.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000242108.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000444777.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000444777.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000444777.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000468591.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000468591.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000468591.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000468591.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000468591.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000468591.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000487069.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000487069.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000487069.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000487069.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000487069.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000487069.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000534978.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000534978.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000534978.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000534978.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000534978.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000534978.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000534978.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000534978.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000534978.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000534978.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000534978.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000534978.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
ENST00000534978.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
NM_030636.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
NM_030636.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
NM_030636.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
NM_030636.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
NM_030636.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
NM_030636.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
NM_030636.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
NM_030636.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
NM_030636.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
NM_030636.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
NM_030636.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
NM_030636.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
NM_030636.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
NM_030636.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
NM_030636.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700007191.hg.1 |
NM_030636.2 |
Refseq |