| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000345117.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000345117.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000345117.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000345117.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000345117.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000345117.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000345117.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000345117.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000529779.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000529779.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000529779.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000529779.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000532093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
ENST00000532093.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
NM_199003.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
NM_199003.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
NM_199003.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
NM_199003.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
NM_199003.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
NM_199003.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
NM_199003.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010298.hg.1 |
NM_199003.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000254250.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000345117.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000345117.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000345117.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000345117.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000345117.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000345117.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000345117.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000345117.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000529779.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000529779.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000529779.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000529779.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000532093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
ENST00000532093.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
NM_018105.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
NM_199003.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
NM_199003.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
NM_199003.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
NM_199003.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
NM_199003.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
NM_199003.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
NM_199003.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800010298.hg.1 |
NM_199003.1 |
Refseq |