Search
Search
| Gene Symbol: FRY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023037.2 | NP_075463.2 | 38 - 39 | 5512 | 71 | |
| XM_006719749.3 | XP_006719812.1 | 38 - 39 | 6593 | 71 | ||
| XM_017020303.1 | XP_016875792.1 | 38 - 39 | 7070 | 71 | ||
| XM_017020304.1 | XP_016875793.1 | 38 - 39 | 5512 | 71 | ||
| XM_017020305.1 | XP_016875794.1 | 38 - 39 | 7089 | 71 | ||
| XM_017020306.1 | XP_016875795.1 | 17 - 18 | 2231 | 71 | ||
| XM_017020307.1 | XP_016875796.1 | 11 - 12 | 1584 | 71 | ||
| GenBank mRNA | AL049784.1 | - | 38 - 39 | 5509 | 71 | |
| U50534.1 | - | 38 - 39 | 5511 | 71 | ||
Set Membership: |
|