| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007051.hg.1 |
ENST00000440955.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007051.hg.1 |
ENST00000440955.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007051.hg.1 |
ENST00000440955.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007051.hg.1 |
ENST00000440955.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007051.hg.1 |
ENST00000440955.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007051.hg.1 |
ENST00000440955.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007051.hg.1 |
NR_110387.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007051.hg.1 |
NR_110387.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007051.hg.1 |
NR_110387.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007051.hg.1 |
NR_110387.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007051.hg.1 |
NR_110387.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007051.hg.1 |
NR_110387.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00147988.hg.1 |
ENST00000440955.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00147988.hg.1 |
NR_110387.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00147988.hg.1 |
ENST00000440955.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00147988.hg.1 |
NR_110387.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00147988.hg.1 |
TC0X00007051.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00147989.hg.1 |
ENST00000440955.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00147989.hg.1 |
NR_110387.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00147989.hg.1 |
ENST00000440955.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00147989.hg.1 |
NR_110387.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00147989.hg.1 |
TC0X00007051.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00007051.hg.1 |
ENST00000440955.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007051.hg.1 |
ENST00000440955.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007051.hg.1 |
ENST00000440955.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007051.hg.1 |
ENST00000440955.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007051.hg.1 |
ENST00000440955.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007051.hg.1 |
ENST00000440955.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007051.hg.1 |
NR_110387.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007051.hg.1 |
NR_110387.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007051.hg.1 |
NR_110387.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007051.hg.1 |
NR_110387.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007051.hg.1 |
NR_110387.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007051.hg.1 |
NR_110387.1 |
Refseq |