| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000543094.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000543094.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000543094.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000543094.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000543094.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000546001.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000546001.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000546001.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000546001.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
ENST00000546001.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12022449.hg.1 |
ENST00000286574.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12022449.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12022449.hg.1 |
TC12001681.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12022450.hg.1 |
ENST00000286574.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12022450.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12022450.hg.1 |
TC12001681.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000543094.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000543094.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000543094.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000543094.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000543094.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000546001.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000546001.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000546001.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000546001.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
ENST00000546001.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200011108.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200186276.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200186276.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200186276.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200186276.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200186276.hg.1 |
TC1200011108.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200186278.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200186278.hg.1 |
ENST00000546001.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200186278.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200186278.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200186278.hg.1 |
ENST00000546001.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200186278.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200186278.hg.1 |
TC1200011108.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200186277.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200186277.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200186277.hg.1 |
ENST00000286574.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200186277.hg.1 |
NM_007191.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200186277.hg.1 |
TC1200011108.hg.1 |
Clariom D TC |