| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000564336.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000564336.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000564336.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000564336.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000564336.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000564336.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000566396.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000566396.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000566396.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000566396.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000566396.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
ENST00000566396.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16006095.hg.1 |
ENST00000268459.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16006095.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16006095.hg.1 |
TC16000440.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1600156957.hg.1 |
ENST00000268459.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600156957.hg.1 |
ENST00000566396.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600156957.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600156957.hg.1 |
TC1600007819.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600156957.hg.1 |
ENST00000268459.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600156957.hg.1 |
ENST00000566396.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600156957.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000268459.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000564336.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000564336.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000564336.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000564336.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000564336.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000564336.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000566396.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000566396.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000566396.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000566396.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000566396.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
ENST00000566396.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600007819.hg.1 |
NM_033119.4 |
Refseq |