| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X001981.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X001981.hg.1 |
ENST00000536160.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X001981.hg.1 |
ENST00000545017.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X001981.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X001981.hg.1 |
TC0X000171.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
ENST00000492288.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
ENST00000492288.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
ENST00000492288.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
ENST00000492288.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006958.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00147415.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00147415.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00147415.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00147415.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00147415.hg.1 |
TC0X00006958.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
ENST00000378588.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
ENST00000492288.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
ENST00000492288.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
ENST00000492288.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
ENST00000492288.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
NM_000397.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00006958.hg.1 |
NM_000397.3 |
Refseq |