| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Clariom D Assay |
PSR0700169192.hg.1 |
ENST00000397238.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700169192.hg.1 |
ENST00000461065.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700169192.hg.1 |
ENST00000463381.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700169192.hg.1 |
ENST00000473633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700169192.hg.1 |
ENST00000622464.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700169192.hg.1 |
NM_001281300.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700169192.hg.1 |
NM_031946.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700169192.hg.1 |
ENST00000397238.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700169192.hg.1 |
ENST00000461065.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700169192.hg.1 |
ENST00000463381.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700169192.hg.1 |
ENST00000473633.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700169192.hg.1 |
ENST00000622464.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700169192.hg.1 |
NM_001281300.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700169192.hg.1 |
NM_031946.6 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700169192.hg.1 |
TC0700009693.hg.1 |
Clariom D TC |