| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
ENST00000266659.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
ENST00000266659.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
ENST00000266659.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
ENST00000266659.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
ENST00000266659.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
ENST00000266659.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
ENST00000266659.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
ENST00000266659.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
ENST00000266659.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
ENST00000266659.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
ENST00000266659.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
ENST00000266659.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
ENST00000456650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
ENST00000456650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
ENST00000456650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
ENST00000456650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
ENST00000456650.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
ENST00000536703.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
ENST00000536703.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
ENST00000536703.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
ENST00000536703.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
ENST00000536703.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
ENST00000536703.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
ENST00000536703.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
ENST00000536703.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
ENST00000536703.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
ENST00000550491.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
ENST00000550491.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
ENST00000550491.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
ENST00000550491.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
NM_006851.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
NM_006851.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
NM_006851.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
NM_006851.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
NM_006851.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
NM_006851.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
NM_006851.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
NM_006851.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
NM_006851.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200008224.hg.1 |
NM_006851.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
ENST00000266659.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
ENST00000266659.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
ENST00000266659.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
ENST00000266659.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
ENST00000266659.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
ENST00000266659.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
ENST00000266659.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
ENST00000266659.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
ENST00000266659.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
ENST00000266659.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
ENST00000266659.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
ENST00000266659.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
ENST00000456650.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
ENST00000456650.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
ENST00000456650.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
ENST00000456650.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
ENST00000456650.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
ENST00000536703.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
ENST00000536703.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
ENST00000536703.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
ENST00000536703.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
ENST00000536703.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
ENST00000536703.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
ENST00000536703.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
ENST00000536703.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
ENST00000536703.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
ENST00000550491.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
ENST00000550491.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
ENST00000550491.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
ENST00000550491.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
NM_006851.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
NM_006851.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
NM_006851.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
NM_006851.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
NM_006851.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
NM_006851.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
NM_006851.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
NM_006851.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
NM_006851.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200008224.hg.1 |
NM_006851.2 |
Refseq |