| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02029146.hg.1 |
ENST00000294947.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02029146.hg.1 |
ENST00000422269.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02029146.hg.1 |
ENST00000445927.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02029146.hg.1 |
NM_001163561.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02029146.hg.1 |
NM_173649.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02029146.hg.1 |
TC02005061.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
ENST00000294947.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
ENST00000294947.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
ENST00000294947.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
ENST00000294947.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
ENST00000294947.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
ENST00000294947.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
ENST00000294947.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
ENST00000294947.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
ENST00000445927.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
ENST00000445927.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
ENST00000445927.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
ENST00000445927.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
ENST00000445927.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
ENST00000445927.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
ENST00000445927.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
ENST00000445927.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
ENST00000445927.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
ENST00000445927.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
ENST00000449846.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
ENST00000449846.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
ENST00000449846.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
ENST00000449846.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
ENST00000449846.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
NM_001163561.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
NM_001163561.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
NM_001163561.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
NM_001163561.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
NM_001163561.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
NM_001163561.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
NM_001163561.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
NM_001163561.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
NM_001163561.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
NM_001163561.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
NM_173649.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
NM_173649.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
NM_173649.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
NM_173649.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
NM_173649.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
NM_173649.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
NM_173649.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
NM_173649.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
NM_173649.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200016661.hg.1 |
NM_173649.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
ENST00000294947.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
ENST00000294947.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
ENST00000294947.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
ENST00000294947.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
ENST00000294947.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
ENST00000294947.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
ENST00000294947.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
ENST00000294947.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
ENST00000445927.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
ENST00000445927.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
ENST00000445927.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
ENST00000445927.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
ENST00000445927.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
ENST00000445927.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
ENST00000445927.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
ENST00000445927.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
ENST00000445927.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
ENST00000445927.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
ENST00000449846.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
ENST00000449846.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
ENST00000449846.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
ENST00000449846.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
ENST00000449846.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
NM_001163561.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
NM_001163561.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
NM_001163561.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
NM_001163561.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
NM_001163561.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
NM_001163561.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
NM_001163561.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
NM_001163561.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
NM_001163561.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
NM_001163561.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
NM_173649.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
NM_173649.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
NM_173649.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
NM_173649.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
NM_173649.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
NM_173649.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
NM_173649.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
NM_173649.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
NM_173649.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200016661.hg.1 |
NM_173649.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200226200.hg.1 |
ENST00000294947.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200226200.hg.1 |
ENST00000445927.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200226200.hg.1 |
NM_001163561.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200226200.hg.1 |
NM_173649.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200226200.hg.1 |
ENST00000294947.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200226200.hg.1 |
ENST00000445927.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200226200.hg.1 |
NM_001163561.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200226200.hg.1 |
NM_173649.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200226200.hg.1 |
TC0200016661.hg.1 |
Clariom D TC |