| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002150.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002150.hg.1 |
TC6_dbb_hap3000118.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002156.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002156.hg.1 |
TC6_dbb_hap3000118.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001937.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001937.hg.1 |
TC6_mcf_hap5000109.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001947.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001947.hg.1 |
TC6_mcf_hap5000109.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002194.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002194.hg.1 |
TC6_qbl_hap6000119.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002195.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002195.hg.1 |
TC6_qbl_hap6000119.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7001994.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7001994.hg.1 |
TC6_ssto_hap7000110.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06016759.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06016759.hg.1 |
TC06001492.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000892.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000892.hg.1 |
TC6_apd_hap1000065.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002219.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002219.hg.1 |
TC6_cox_hap2000129.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002226.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002226.hg.1 |
TC6_cox_hap2000129.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001739.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001739.hg.1 |
TC6_mann_hap4000107.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001746.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001746.hg.1 |
TC6_mann_hap4000107.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001940.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001940.hg.1 |
TC6_mcf_hap5000109.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001941.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001941.hg.1 |
TC6_mcf_hap5000109.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06016751.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06016751.hg.1 |
TC06001492.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06016755.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06016755.hg.1 |
TC06001492.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002199.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002199.hg.1 |
TC6_qbl_hap6000119.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7001993.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7001993.hg.1 |
TC6_ssto_hap7000110.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002000.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002000.hg.1 |
TC6_ssto_hap7000110.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06016754.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06016754.hg.1 |
TC06001492.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000893.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000893.hg.1 |
TC6_apd_hap1000065.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000899.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000899.hg.1 |
TC6_apd_hap1000065.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002146.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002146.hg.1 |
TC6_dbb_hap3000118.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000889.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000889.hg.1 |
TC6_apd_hap1000065.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002154.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002154.hg.1 |
TC6_dbb_hap3000118.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001736.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001736.hg.1 |
TC6_mann_hap4000107.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001945.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001945.hg.1 |
TC6_mcf_hap5000109.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7001990.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7001990.hg.1 |
TC6_ssto_hap7000110.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7001998.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7001998.hg.1 |
TC6_ssto_hap7000110.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002224.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002224.hg.1 |
TC6_cox_hap2000129.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002191.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002191.hg.1 |
TC6_qbl_hap6000119.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06016761.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06016761.hg.1 |
TC06001492.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002216.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002216.hg.1 |
TC6_cox_hap2000129.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001740.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001740.hg.1 |
TC6_mann_hap4000107.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002201.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002201.hg.1 |
TC6_qbl_hap6000119.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000897.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000897.hg.1 |
TC6_apd_hap1000065.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002220.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002220.hg.1 |
TC6_cox_hap2000129.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002149.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002149.hg.1 |
TC6_dbb_hap3000118.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001744.hg.1 |
NR_026751.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001744.hg.1 |
TC6_mann_hap4000107.hg.1 |
HTA TC |