| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02043528.hg.1 |
ENST00000452335.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02043528.hg.1 |
ENST00000477734.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02043528.hg.1 |
NM_001093730.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02043528.hg.1 |
TC02002701.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
ENST00000477734.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
ENST00000477734.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
ENST00000477734.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
ENST00000477734.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
ENST00000452335.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
ENST00000477734.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
ENST00000477734.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
ENST00000477734.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
ENST00000477734.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200015525.hg.1 |
NM_001093730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200213983.hg.1 |
ENST00000452335.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200213983.hg.1 |
ENST00000477734.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200213983.hg.1 |
NM_001093730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200213983.hg.1 |
ENST00000452335.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200213983.hg.1 |
ENST00000477734.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200213983.hg.1 |
NM_001093730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200213983.hg.1 |
TC0200015525.hg.1 |
Clariom D TC |