Search
Search

C11orf95| Gene Symbol: C11orf95 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB231750.1 | - | 2 | 250 | 115 | |
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | reticulon 3 | RTN3 |
ASYIP, HAP, NSPL2, NSPLII, RTN3-A1 |
-3.9kb | Anti-sense | Chr. 11 - 63681315 - 63759891 on NCBI Build GRCh38 |
| Downstream | chromosome 11 open reading frame 84 | C11orf84 | 49.3kb | Anti-sense | Chr. 11 - 63813374 - 63827718 on NCBI Build GRCh38 |
Set Membership: |
|
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000338498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000338498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000338498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000338498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000338498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000338498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000338498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000338498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000338498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000338498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000338498.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000458348.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000458348.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000458348.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000458348.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000458348.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000546282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000546282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000546282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000546282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000546282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000546282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000546282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000546282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000546282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | ENST00000546282.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000338498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000338498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000338498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000338498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000338498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000338498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000338498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000338498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000338498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000338498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000338498.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000433688.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000445014.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000458348.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000458348.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000458348.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000458348.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000458348.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000546282.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000546282.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000546282.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000546282.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000546282.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000546282.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000546282.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000546282.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000546282.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | ENST00000546282.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | NM_001144936.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100011131.hg.1 | NM_001144936.1 | Refseq |