| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19022038.hg.1 |
ENST00000006275.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19022038.hg.1 |
NM_024108.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19022038.hg.1 |
TC19001615.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000006275.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000006275.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000006275.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000006275.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000006275.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000006275.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000006275.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000006275.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000006275.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000006275.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000585934.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000585934.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000585934.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000585934.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000585934.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000585934.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000585934.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000585934.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000585934.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000587818.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000587818.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000587818.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000587818.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000587818.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000588062.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000588062.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000588062.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000588062.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000588062.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000588062.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000588062.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000588062.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000592647.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000592647.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000592647.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000592647.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000592647.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000592647.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000592647.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
ENST00000592647.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_001270891.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_001270891.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_001270891.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_001270891.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_001270891.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_001270891.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_001270891.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_001270891.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_001270891.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_001270892.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_001270892.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_001270892.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_001270892.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_001270892.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_001270892.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_001270892.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_001270892.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_001270892.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_001270893.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_001270893.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_001270893.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_001270893.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_001270893.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_001270893.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_001270893.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_001270893.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_024108.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_024108.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_024108.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_024108.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_024108.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_024108.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_024108.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_024108.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_024108.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010926.hg.1 |
NM_024108.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900190655.hg.1 |
ENST00000006275.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900190655.hg.1 |
ENST00000585934.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900190655.hg.1 |
ENST00000588062.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900190655.hg.1 |
ENST00000592647.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900190655.hg.1 |
NM_001270891.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900190655.hg.1 |
NM_001270892.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900190655.hg.1 |
NM_001270893.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900190655.hg.1 |
NM_024108.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900190655.hg.1 |
TC1900010926.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900190655.hg.1 |
ENST00000006275.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900190655.hg.1 |
ENST00000585934.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900190655.hg.1 |
ENST00000588062.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900190655.hg.1 |
ENST00000592647.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900190655.hg.1 |
NM_001270891.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900190655.hg.1 |
NM_001270892.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900190655.hg.1 |
NM_001270893.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900190655.hg.1 |
NM_024108.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000006275.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000006275.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000006275.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000006275.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000006275.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000006275.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000006275.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000006275.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000006275.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000006275.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000585934.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000585934.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000585934.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000585934.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000585934.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000585934.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000585934.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000585934.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000585934.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000587818.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000587818.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000587818.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000587818.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000587818.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000588062.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000588062.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000588062.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000588062.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000588062.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000588062.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000588062.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000588062.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000592647.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000592647.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000592647.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000592647.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000592647.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000592647.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000592647.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
ENST00000592647.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_001270891.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_001270891.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_001270891.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_001270891.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_001270891.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_001270891.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_001270891.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_001270891.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_001270891.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_001270892.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_001270892.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_001270892.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_001270892.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_001270892.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_001270892.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_001270892.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_001270892.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_001270892.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_001270893.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_001270893.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_001270893.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_001270893.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_001270893.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_001270893.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_001270893.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_001270893.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_024108.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_024108.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_024108.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_024108.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_024108.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_024108.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_024108.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_024108.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_024108.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010926.hg.1 |
NM_024108.2 |
Refseq |