| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
ENST00000468296.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
ENST00000468296.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
ENST00000468296.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
ENST00000468296.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
ENST00000468296.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
ENST00000468296.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
ENST00000468296.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
ENST00000471541.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
ENST00000471541.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
ENST00000471541.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
ENST00000471541.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
ENST00000471541.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
ENST00000471541.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
ENST00000477374.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
ENST00000477374.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
ENST00000477374.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
ENST00000477374.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
ENST00000477374.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
ENST00000477374.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
ENST00000477374.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
ENST00000478296.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
ENST00000478296.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
ENST00000478296.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
ENST00000478296.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
ENST00000478296.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
ENST00000478296.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
ENST00000478296.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
ENST00000491507.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
ENST00000491507.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
ENST00000491507.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
ENST00000491507.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
ENST00000491507.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
ENST00000491507.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
ENST00000491507.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
NM_001128223.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
NM_001128223.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
NM_001128223.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
NM_001128223.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
NM_001128223.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
NM_001128223.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
NM_001128223.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
NM_001128223.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
NM_001128223.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
NM_001290208.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
NM_001290208.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
NM_001290208.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
NM_001290208.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
NM_001290208.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
NM_001290208.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
NM_001290208.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
NM_001290209.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
NM_001290209.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
NM_001290209.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
NM_001290209.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
NM_001290209.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
NM_001290209.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
NM_001290209.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
NM_001290210.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
NM_001290210.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
NM_001290210.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
NM_001290210.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
NM_001290210.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
NM_001290210.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011597.hg.1 |
NM_001290210.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300186614.hg.1 |
ENST00000477374.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300186614.hg.1 |
NM_001290210.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300186614.hg.1 |
ENST00000477374.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300186614.hg.1 |
NM_001290210.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300186614.hg.1 |
TC0300011597.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
ENST00000468296.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
ENST00000468296.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
ENST00000468296.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
ENST00000468296.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
ENST00000468296.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
ENST00000468296.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
ENST00000468296.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
ENST00000471541.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
ENST00000471541.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
ENST00000471541.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
ENST00000471541.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
ENST00000471541.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
ENST00000471541.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
ENST00000477374.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
ENST00000477374.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
ENST00000477374.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
ENST00000477374.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
ENST00000477374.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
ENST00000477374.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
ENST00000477374.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
ENST00000478296.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
ENST00000478296.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
ENST00000478296.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
ENST00000478296.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
ENST00000478296.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
ENST00000478296.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
ENST00000478296.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
ENST00000491507.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
ENST00000491507.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
ENST00000491507.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
ENST00000491507.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
ENST00000491507.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
ENST00000491507.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
ENST00000491507.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
NM_001128223.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
NM_001128223.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
NM_001128223.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
NM_001128223.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
NM_001128223.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
NM_001128223.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
NM_001128223.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
NM_001128223.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
NM_001128223.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
NM_001290208.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
NM_001290208.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
NM_001290208.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
NM_001290208.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
NM_001290208.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
NM_001290208.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
NM_001290208.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
NM_001290209.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
NM_001290209.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
NM_001290209.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
NM_001290209.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
NM_001290209.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
NM_001290209.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
NM_001290209.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
NM_001290210.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
NM_001290210.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
NM_001290210.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
NM_001290210.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
NM_001290210.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
NM_001290210.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011597.hg.1 |
NM_001290210.1 |
Refseq |