| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007075.hg.1 |
ENST00000408964.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007075.hg.1 |
ENST00000408964.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007075.hg.1 |
ENST00000408964.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007075.hg.1 |
ENST00000408964.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007075.hg.1 |
NM_032266.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007075.hg.1 |
NM_032266.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007075.hg.1 |
NM_032266.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200007075.hg.1 |
NM_032266.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007075.hg.1 |
ENST00000408964.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007075.hg.1 |
ENST00000408964.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007075.hg.1 |
ENST00000408964.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007075.hg.1 |
ENST00000408964.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200007075.hg.1 |
NM_032266.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007075.hg.1 |
NM_032266.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007075.hg.1 |
NM_032266.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200007075.hg.1 |
NM_032266.3 |
Refseq |