Search
Search

KRBA2| Gene Symbol: KRBA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213597.3 | NP_998762.1 | 1 - 2 | 230 | 67 | |
| XM_005256443.2 | XP_005256500.1 | 4 - 5 | 1387 | 67 | ||
| XM_011523643.1 | XP_011521945.1 | 3 - 4 | 1203 | 67 | ||
| XM_011523644.1 | XP_011521946.1 | 2 - 3 | 994 | 67 | ||
| XM_017024163.1 | XP_016879652.1 | 3 - 4 | 1476 | 67 | ||
| GenBank mRNA | AK131219.1 | - | 1 - 2 | 231 | 67 | |
| BC033390.1 | - | 1 - 2 | 65 | 67 | ||
Set Membership: |
|