Search
Search

SFPQ| Gene Symbol: SFPQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005544006.2 | XP_005544063.1 | 5 - 6 | 1697 | 140 | |
| XM_015438579.1 | XP_015294065.1 | 5 - 6 | 1697 | 140 | ||
| XM_015438587.1 | XP_015294073.1 | 5 - 6 | 1697 | 140 | ||
| XM_015438596.1 | XP_015294082.1 | 5 - 6 | 1697 | 140 | ||
| XM_015438602.1 | XP_015294088.1 | 5 - 6 | 1697 | 140 | ||
| XM_015438605.1 | XP_015294091.1 | 5 - 6 | 1937 | 140 | ||
| XM_015438607.1 | XP_015294093.1 | 5 - 6 | 1937 | 140 | ||
| GenBank mRNA | AB179137.1 | - | 4 - 5 | 652 | 140 | |
Set Membership: |
|