Search
Search

JMJD1C| Gene Symbol: JMJD1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005565742.2 | XP_005565799.1 | 2 - 3 | 770 | 96 | |
| XM_005565743.2 | XP_005565800.1 | 2 - 3 | 770 | 96 | ||
| XM_005565746.2 | XP_005565803.1 | 2 - 3 | 267 | 96 | ||
| XM_015456292.1 | XP_015311778.1 | 3 - 4 | 350 | 96 | ||
Set Membership: |
|