Search
Search

THRAP3| Gene Symbol: THRAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005543957.2 | XP_005544014.1 | 3 - 4 | 1725 | 146 | |
| XM_005543958.2 | XP_005544015.1 | 3 - 4 | 1728 | 146 | ||
Set Membership: |
|