Search
Search

RNF212B| Gene Symbol: RNF212B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005560850.2 | XP_005560907.1 | 4 - 5 | 405 | 81 | |
| XM_005560851.2 | XP_005560908.1 | 4 - 5 | 405 | 81 | ||
Set Membership: |
|