Search
Search

SPATA5| Gene Symbol: SPATA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005555872.2 | XP_005555929.1 | 15 - 16 | 2575 | 133 | |
| XM_005555873.2 | XP_005555930.1 | 14 - 15 | 2233 | 133 | ||
| XM_015450802.1 | XP_015306288.1 | 15 - 16 | 2572 | 133 | ||
Set Membership: |
|