Search
Search

ATP12A| Gene Symbol: ATP12A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005585490.2 | XP_005585547.1 | 8 - 9 | 1370 | 98 | |
| XM_005585491.2 | XP_005585548.1 | 8 - 9 | 1251 | 98 | ||
Set Membership: |
|