Search
Search

NDRG4| Gene Symbol: NDRG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005592101.2 | XP_005592158.1 | 16 - 17 | 1525 | 79 | |
| XM_005592104.2 | XP_005592161.1 | 16 - 17 | 1167 | 79 | ||
| XM_005592105.2 | XP_005592162.1 | 15 - 16 | 1435 | 79 | ||
| XM_005592107.2 | XP_005592164.1 | 15 - 16 | 1262 | 79 | ||
| XM_005592109.2 | XP_005592166.1 | 14 - 15 | 1390 | 79 | ||
| XM_005592110.2 | XP_005592167.1 | 15 - 16 | 1157 | 79 | ||
| XM_005592111.2 | XP_005592168.1 | 14 - 15 | 1167 | 79 | ||
| XM_005592115.2 | XP_005592172.1 | 13 - 14 | 1069 | 79 | ||
| XM_015442808.1 | XP_015298294.1 | 14 - 15 | 1058 | 79 | ||
Set Membership: |
|