Search
Search

PLEKHO1| Gene Symbol: PLEKHO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005542028.2 | XP_005542085.2 | 2 - 3 | 460 | 114 | |
| XM_005542031.2 | XP_005542088.1 | 2 - 3 | 466 | 114 | ||
| XM_015457846.1 | XP_015313332.1 | 2 - 3 | 713 | 114 | ||
| XM_015457850.1 | XP_015313336.1 | 2 - 3 | 713 | 114 | ||
| XM_015457855.1 | XP_015313341.1 | 2 - 3 | 713 | 114 | ||
| XM_015457859.1 | XP_015313345.1 | 2 - 3 | 713 | 114 | ||
| XM_015457865.1 | XP_015313351.1 | 2 - 3 | 713 | 114 | ||
Set Membership: |
|