Search
Search

NAA35| Gene Symbol: NAA35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287681.1 | NP_001274610.1 | 17 - 18 | 1694 | 102 | |
| XM_005582057.2 | XP_005582114.1 | 17 - 18 | 1715 | 102 | ||
| XM_005582058.2 | XP_005582115.1 | 17 - 18 | 1713 | 102 | ||
| XM_005582059.2 | XP_005582116.1 | 17 - 18 | 1917 | 102 | ||
| XM_005582060.2 | XP_005582117.1 | 17 - 18 | 1777 | 102 | ||
| XM_005582061.2 | XP_005582118.1 | 17 - 18 | 1685 | 102 | ||
| GenBank mRNA | AB169022.1 | - | 17 - 18 | 1694 | 102 | |
Set Membership: |
|