Search
Search

SYT4| Gene Symbol: SYT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005586819.2 | XP_005586876.1 | 3 - 4 | 1339 | 71 | |
| XM_005586820.2 | XP_005586877.1 | 3 - 4 | 1331 | 71 | ||
| XM_005586821.2 | XP_005586878.1 | 3 - 4 | 1328 | 71 | ||
Set Membership: |
|