Search
Search

ZCCHC4| Gene Symbol: ZCCHC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005554608.2 | XP_005554665.1 | 1 - 2 | 161 | 93 | |
| XM_005554609.2 | XP_005554666.1 | 1 - 2 | 161 | 93 | ||
| XM_005554610.2 | XP_005554667.1 | 1 - 2 | 161 | 93 | ||
| XM_015450218.1 | XP_015305704.1 | 1 - 2 | 161 | 93 | ||
Set Membership: |
|