Search
Search

RDH11| Gene Symbol: RDH11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283857.1 | NP_001270786.1 | 1 - 2 | 102 | 86 | |
| XM_005561572.2 | XP_005561629.1 | 1 - 2 | 335 | 86 | ||
| XM_005561574.2 | XP_005561631.1 | 1 - 2 | 361 | 86 | ||
| GenBank mRNA | AB170555.1 | - | 1 - 2 | 107 | 86 | |
| AB170891.1 | - | 1 - 2 | 102 | 86 | ||
| DQ078725.1 | - | 1 - 2 | 75 | 86 | ||
Set Membership: |
|