Search
Search

CCDC33| Gene Symbol: CCDC33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005560060.2 | XP_005560117.1 | 8 - 9 | 1311 | 95 | |
| XM_005560061.2 | XP_005560118.1 | 8 - 9 | 1311 | 95 | ||
| XM_005560062.2 | XP_005560119.1 | 8 - 9 | 1311 | 95 | ||
| XM_015453073.1 | XP_015308559.1 | 8 - 9 | 1311 | 95 | ||
| XM_015453074.1 | XP_015308560.1 | 8 - 9 | 1194 | 95 | ||
| GenBank mRNA | AB070039.1 | - | 8 - 9 | 1276 | 95 | |
Set Membership: |
|