Search
Search

DIXDC1| Gene Symbol: DIXDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005579605.2 | XP_005579662.1 | 2 - 3 | 488 | 65 | |
| XM_005579607.2 | XP_005579664.1 | 3 - 4 | 329 | 65 | ||
| XM_005579608.2 | XP_005579665.1 | 3 - 4 | 552 | 65 | ||
| XM_015435703.1 | XP_015291189.1 | 3 - 4 | 324 | 65 | ||
| XM_015435704.1 | XP_015291190.1 | 3 - 4 | 465 | 65 | ||
| XM_015435705.1 | XP_015291191.1 | 2 - 3 | 207 | 65 | ||
Set Membership: |
|