Search
Search

ABCC12| Gene Symbol: ABCC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015443238.1 | XP_015298724.1 | 18 - 19 | 2565 | 123 | |
| XM_015443240.1 | XP_015298726.1 | 17 - 18 | 2511 | 123 | ||
| XM_015443243.1 | XP_015298729.1 | 18 - 19 | 2565 | 123 | ||
Set Membership: |
|