Search
Search

RBM5| Gene Symbol: RBM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005547174.2 | XP_005547231.1 | 15 - 16 | 1452 | 77 | |
| XM_005547175.2 | XP_005547232.1 | 15 - 16 | 1528 | 77 | ||
| XM_005547177.2 | XP_005547234.1 | 15 - 16 | 1453 | 77 | ||
| GenBank mRNA | AB170932.1 | - | 15 - 16 | 1426 | 77 | |
Set Membership: |
|